Methyl acetate ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 79-20-9 Molecular Formula: CH3COOCH3 Formula Weight: 74.08 Flash Point: -10°(14°F) Refractive Index: 1.3620 Odour: Pleasant Melting Point: -98° Boiling Point: 56-58° Beilstein Reference: 1736662 Merck Reference: 14,6008 UN No.: 1231 MDL No.: MFCD00008711
Synonyms: Acetic acid methyl ester
- CAS Number:79-20-9
- Formula:C₃H₆O₂
- MDL Number:MFCD00008711
- Molecular Weight:74.08 g/mol
- Merck Index:14,6008
- Boiling Pt:56…58 °C
- Melting Pt:–98 °C
- Density:0.934
- Flash Pt:–10 °C (14 °F)
- Storage Temperature:Ambient
- EINECS:201-185-2
- UN:1231
- ADR:3,II
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 79-20-9 Molecular Formula: CH3COOCH3 Formula Weight: 74.08 Flash Point: -10°(14°F) Refractive Index: 1.3620 Odour: Pleasant Melting Point: -98° Boiling Point: 56-58° Beilstein Reference: 1736662 Merck Reference: 14,6008 UN No.: 1231 MDL No.: MFCD00008711
Synonyms: Acetic acid methyl ester
- CAS Number:79-20-9
- Formula:C₃H₆O₂
- MDL Number:MFCD00008711
- Molecular Weight:74.08 g/mol
- Merck Index:14,6008
- Boiling Pt:56…58 °C
- Melting Pt:–98 °C
- Density:0.934
- Flash Pt:–10 °C (14 °F)
- Storage Temperature:Ambient
- EINECS:201-185-2
- UN:1231
- ADR:3,II



