Order Entry
Export
Orders LinkContactUsLinkComponent
Ethyl acetate

Ethyl acetate

Flammable
Irritant
   Sustainable Options Available
Supplier:   Roth Carl
Ethyl acetate
Supplier:  Roth Carl
   Sustainable Options Available
Certificates
Ethyl acetate

3 Options Available Below

Product Details & Documents
Synonyms:  Acetic acid ethyl ester
  • CAS Number:141-78-6
  • Formula:CH₃COOC₂H₅
  • MDL Number:MFCD00009171
  • Molecular Weight:88.11 g/mol
  • Merck Index:13,03792
  • Boiling Pt:77.1 °C (1013 hPa)
  • Melting Pt:–83 °C
  • Density:0.902 g/cm³ (20 °C)
  • Flash Pt:–4 °C
  • Storage Temperature:Ambient
  • EINECS:205-500-4
  • UN:1173
  • ADR:3,II
  • REACH:01-2119475103-46
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ROTH7338.1
  • ROTH7338.3
  • ROTH7338.2
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • Chem21 Solvent Selection Guide recommended by default and after discussion
  • Chem21 Solvent Selection Guide recommended by default and after discussion
  • Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 33,20 €
    33,20 €
    1 * 1 L
       33,20 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    ROTH7338.1
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended by default and after discussion
      Safer Human and Environmental Health
  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 56,00 €
    56,00 €
    1 * 2,5 L
       56,00 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    ROTH7338.3
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended by default and after discussion
      Safer Human and Environmental Health
  • Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 151,00 €
    151,00 €
    1 * 10 L
       151,00 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    ROTH7338.2
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended by default and after discussion
      Safer Human and Environmental Health