Ethyl acetate
Supplier: Roth Carl
About this item
Synonyms: Acetic acid ethyl ester
- CAS Number:141-78-6
- Formula:CH₃COOC₂H₅
- MDL Number:MFCD00009171
- Molecular Weight:88.11 g/mol
- Merck Index:13,03792
- Boiling Pt:77.1 °C (1013 hPa)
- Melting Pt:–83 °C
- Density:0.902 g/cm³ (20 °C)
- Flash Pt:–4 °C
- Storage Temperature:Ambient
- EINECS:205-500-4
- UN:1173
- ADR:3,II
- REACH:01-2119475103-46
5 Options Available Below
Product Details & Documents
Synonyms: Acetic acid ethyl ester
- CAS Number:141-78-6
- Formula:CH₃COOC₂H₅
- MDL Number:MFCD00009171
- Molecular Weight:88.11 g/mol
- Merck Index:13,03792
- Boiling Pt:77.1 °C (1013 hPa)
- Melting Pt:–83 °C
- Density:0.902 g/cm³ (20 °C)
- Flash Pt:–4 °C
- Storage Temperature:Ambient
- EINECS:205-500-4
- UN:1173
- ADR:3,II
- REACH:01-2119475103-46
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- ROTH6784.1
- ROTH6784.3
- ROTH6784.2
- ROTH6784.4
- ROTH6784.5
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- Chem21 Solvent Selection Guide recommended by default and after discussion
- Chem21 Solvent Selection Guide recommended by default and after discussion
- Chem21 Solvent Selection Guide recommended by default and after discussion
- Chem21 Solvent Selection Guide recommended by default and after discussion
- Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
Safer Human and Environmental Health More
Specifications
Cat. No.ROTH6784.1Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended by default and after discussionSafer Human and Environmental Health - Specifications:
Safer Human and Environmental Health More
Specifications
Cat. No.ROTH6784.3Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended by default and after discussionSafer Human and Environmental Health - Specifications:
Safer Human and Environmental Health More
Specifications
Cat. No.ROTH6784.2Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended by default and after discussionSafer Human and Environmental Health - Specifications:
Safer Human and Environmental Health More
Specifications
Cat. No.ROTH6784.4Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended by default and after discussionSafer Human and Environmental Health - Specifications:
Safer Human and Environmental Health More
Specifications
Cat. No.ROTH6784.5Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended by default and after discussionSafer Human and Environmental Health



