Ethyl acetate ≥98.5%
Plastic bottle, 1 kg
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: Acetic acid ethyl ester
- CAS Number:141-78-6
- Formula:CH₃COOC₂H₅
- MDL Number:MFCD00009171
- Molecular Weight:88.11 g/mol
- Merck Index:13,03792
- Boiling Pt:77.1 °C (1013 hPa)
- Melting Pt:–83 °C
- Density:0.902 g/cm³ (20 °C)
- Flash Pt:–4 °C
- Storage Temperature:Ambient
- EINECS:205-500-4
- UN:1173
- ADR:3,II
- REACH:01-2119475103-46
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Plastic bottle
- Pk:1 kg



