Order Entry
Export
ContactUsLinkComponent
Isopropyl acetate
Isopropyl acetate
Catalog # PROOCHE161
CAS Number:  
Isopropyl acetate
Catalog # PROOCHE161
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2 mL
  • Seq:
    2145

Specifications

About this item

 :
 :102.13 g/mol
Merck Index:13,05225
Boiling Pt:85 °C (1013 hPa)
Melting Pt:–62 °C
Density:0.8732 g/cm³ (20 °C)
Flash Pt:4 °C
Storage Temperature:Ambient
EINECS:203-561-1
UN:1220
ADR:3,II
 :GHS02
 :GHS07
CAS Number:108-21-4
Formula:C₅H₁₀O₂
MDL Number:MFCD00008877