Isopropyl acetate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108-21-4
:
Formula:C₅H₁₀O₂
MDL Number:MFCD00008877
:102.13 g/mol
Merck Index:14,5205
Boiling Pt:87…89 °C
Melting Pt:–73 °C
Density:0.870
Flash Pt:4 °C (39 °F)
UN:1220
ADR:3,II