Isopropyl acetate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108-21-4
:
Formula:C₅H₁₀O₂
MDL Number:MFCD00008877
:102.13 g/mol
Merck Index:13,05225
Boiling Pt:85 °C (1013 hPa)
Melting Pt:–73 °C
Density:0.872 g/cm³
Flash Pt:4 °C
Storage Temperature:Ambient
EINECS:203-561-1
UN:1220
ADR:3,II
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3760 to 1.3780 (20°C, 589 nm)
- GC:≥98.5 %