Order Entry
Export
ContactUsLinkComponent
 
Isoamyl acetate 99+%, pure
 
undefined
Isoamyl acetate 99+%, pure
 

 

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3990 to 1.4030 (20°C, 589 nm)
  • GC:
    ≥98.5 % (sum of isomers)
  • Acid value:
    ≤2 mg KOH/g