2,3-Pentanedione 97%
About this item
Synonyms: Pentane-2,3-dione
- CAS Number:600-14-6
- Formula:C₅H₈O₂
- MDL Number:MFCD00009313
- Molecular Weight:100.12 g/mol
- Boiling Pt:110…112 °C (760 mmHg)
- Melting Pt:–52 °C
- Density:0.958 g/cm³
- Flash Pt:18 °C
- Storage Temperature:Refrigerator
- EINECS:209-984-8
- UN:1224
- ADR:3,II
3 Options Available Below
Synonyms: Pentane-2,3-dione
- CAS Number:600-14-6
- Formula:C₅H₈O₂
- MDL Number:MFCD00009313
- Molecular Weight:100.12 g/mol
- Boiling Pt:110…112 °C (760 mmHg)
- Melting Pt:–52 °C
- Density:0.958 g/cm³
- Flash Pt:18 °C
- Storage Temperature:Refrigerator
- EINECS:209-984-8
- UN:1224
- ADR:3,II
Specifications for Product Family
Specification Test Results
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4020 to 1.4060 (20°C, 589 nm)
- GC:≥96.0 %



