Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Hexanedione
2,3-Hexanedione ≥94%
2,3-Hexanedione ≥94%
:
Thermo Fisher Scientific
2,3-Hexanedione ≥94%
:
Thermo Fisher Scientific
:
3848-24-6
:
:
C₆H₁₀O₂
:
MFCD00009398
:
127…128 °C
:
–30 °C
:
0.937
:
28 °C (82 °F)
:
Ambient
:
223-350-8
:
3,III
:
114.14 g/mol
:
1224
:
GHS02
:
GHS07
msds_ALFAL04669
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years