Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Racks
Draining Racks
Accessories for draining racks
Accessories for draining racks
BRND95111
:
Brand
Accessories for draining racks
BRND95111
:
Brand
Ø×L:
6×120 mm
Description:
Pegs for draining rack, PS, for narrow mouth vessels, test tubes, small volumetric flasks and graduated cylinders
Pk:
11