Order Entry
Export
ContactUsLinkComponent
Chlorotrimethylsilane
Flammable
Toxic
Corrosive
Chlorotrimethylsilane
Catalog # HYPETS-88530
CAS Number:  
Flammable
Toxic
Corrosive
Chlorotrimethylsilane
Catalog # HYPETS-88530
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Boiling Pt:59 °C (1013 hPa)
Melting Pt:–58 °C
Density:0.8576 g/cm³ (20 °C)
Flash Pt:–18 °C
Storage Temperature:Ambient
EINECS:200-900-5
UN:1298
ADR:3,II
 :108.64 g/mol
REACH:01-2119457596-25
 :GHS02
 :GHS06
 :GHS05
CAS Number:75-77-4
Formula:C₃H₉ClSi
MDL Number:MFCD00000502