Order Entry
Export
ContactUsLinkComponent
[2.2]-Paracyclophane ≥97%, Sigma-Aldrich®
[2.2]-Paracyclophane ≥97%, Sigma-Aldrich®
Catalog # SIALP225-5G-A
Supplier:  Merck
CAS Number:  
[2.2]-Paracyclophane ≥97%, Sigma-Aldrich®
Catalog # SIALP225-5G-A
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Boiling Pt:321 °C (1013 hPa)
Density:1.23 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
 :208.3 g/mol
EINECS:216-644-2
CAS Number:1633-22-3
Formula:C₁₆H₁₆
MDL Number:MFCD00003707