Order Entry
Export
ContactUsLinkComponent
Caffeic acid
Caffeic acid
Catalog # EHERC10934700
Supplier:  EHRENSTORFER
CAS Number:  
Caffeic acid
Catalog # EHERC10934700
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :180.16 g/mol
Merck Index:12,01673
Boiling Pt:366 °C (1013 hPa)
Melting Pt:234…237 °C
EINECS:206-361-2
CAS Number:331-39-5
Formula:C₉H₈O₄
MDL Number:MFCD00004392