Order Entry
Export
Orders LinkContactUsLinkComponent
 

Sodium dihydrogen phosphate

Glass bottle for solids, 500 g
Catalog # RC-097
Supplier:   G-Biosciences
Sodium dihydrogen phosphate
Catalog # RC-097
Supplier:  G-Biosciences
CAS Number:  7558-80-7
Sodium dihydrogen phosphate

Glass bottle for solids, 500 g

CAS Number:  7558-80-7
Price
56,10 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Sodium dihydrogenphosphate (granular), Sodium dihydrogen orthophosphate, Monobasic Sodium Phosphate, Sodium phosphate monobasic
  • CAS Number:7558-80-7
  • Formula:NaH₂PO₄
  • MDL Number:MFCD00003527
  • Molecular Weight:119.98 g/mol
  • Merck Index:12,08806
  • Melting Pt:∼650 °C
  • Density:0.9996 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-449-2
  • REACH:01-2119489796-13

 

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications