Methyl methoxyacetate, Sigma-Aldrich®
SIAL149209-100G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
:
:104.11 g/mol
:129 °C (1013 hPa)
:1.0579 g/cm³ (15 °C)
:3,III
:M-Clarity™ quality level = MQ100
:228-539-9
:3272
:6290-49-9
:C₄H₈O₃
:MFCD00008451