
Methyl methoxyacetate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6290-49-9
:
Formula:C₄H₈O₃
MDL Number:MFCD00008451
:104.11 g/mol
Boiling Pt:129…130 °C (760 mmHg)
Density:1.05 g/cm³
Flash Pt:35 °C
Storage Temperature:Ambient
EINECS:228-539-9
UN:3272
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Clear colorless to light brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3954 to 1.3974 (20°C, 589 nm)
- GC:≥98.5 %