Order Entry
Export
Orders LinkContactUsLinkComponent
Magnesium sulphate

Magnesium sulphate

Glass bottle for solids, 100 g
Catalog # M006
Supplier:   G-Biosciences
Magnesium sulphate
Catalog # M006
Supplier:  G-Biosciences
CAS Number:  7487-88-9
Magnesium sulphate
Magnesium sulphate

Glass bottle for solids, 100 g

CAS Number:  7487-88-9
Price
82,40 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  MgSO4, Magnesium sulphate anhydrous
  • CAS Number:7487-88-9
  • Formula:MgSO₄
  • MDL Number:MFCD00011110
  • Molecular Weight:120.37 g/mol
  • Merck Index:12,05731
  • Melting Pt:1127 °C
  • Density:2.65 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-298-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications