Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lithium methoxide
Lithium methoxide, Sigma-Aldrich®
Lithium methoxide, Sigma-Aldrich®
:
Merck
Lithium methoxide, Sigma-Aldrich®
:
Merck
:
865-34-9
:
:
CH₃LiO
:
MFCD00036357
:
37.98 g/mol
:
64.6 °C (1013 hPa)
:
0.865 g/cm³ (20 °C)
:
212-737-7
:
3206
:
4.2,II
:
M-Clarity™ quality level = MQ100
:
GHS02
:
GHS05