Dipotassium hexachloroosmate, Sigma-Aldrich®
Catalog # SIAL209090-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:481.14 g/mol
Melting Pt:600 °C
Density:3.42 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:240-893-6
CAS Number:16871-60-6
Formula:Cl₆K₂Os
MDL Number:MFCD00011371