Pesticide reference standard, Pentachlorophenol 10 µg/ml in methanol, Dr Ehrenstorfer
EHERL15970000ME
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:87-86-5
- Description:Pentachlorophenol 10 µg/ml in methanol
- Formula:C₆HCl₅O
- Description:Pentachlorophenol