




Hydrazine hydrate (N₂H₄ 50 - 60%), Reagent Grade, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10217-52-4
:7803-57-8
Formula:NH₂NH₂·xH₂O
MDL Number:MFCD00149931
:32.05 g/mol (anhy)
Merck Index:13,04790
Boiling Pt:117.2 °C (1013 hPa)
Melting Pt:–51.7 °C
Density:1.03 g/cm³ (20 °C)
EINECS:206-114-9
UN:2030
ADR:8,II
:M-Clarity™ quality level = MQ200
Flash Pt:96 °C (closed cup)
:GHS02
:GHS06
:GHS05
:GHS08
:GHS09