D-(+)-Biotin ≥97.5%, Pierce™
Glass bottle for solids, 1 g
Catalog # PIER29129
Supplier: Thermo Fisher Scientific
D-(+)-Biotin ≥97.5%, Pierce™
Catalog # PIER29129
Supplier: Thermo Fisher Scientific
CAS Number: 58-85-5
Jump to:
Product Details & Documents
Biotin, also known as vitamin H, is a small molecule (MW 244.3) that is present in tiny amounts in all living cells and is critical for a number of biological processes.
The valeric acid side chain of the biotin molecule can be derivatized in order to incorporate various reactive groups that are used to attach biotin to other molecules. In the context of immunohistochemistry (IHC), biotin is conjugated to antibodies or to the enzyme reporters used to detect target antigens.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Synonyms: Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
- CAS Number:58-85-5
- Formula:C₁₀H₁₆N₂O₃S
- MDL Number:MFCD00005541
- Molecular Weight:244.31 g/mol
- Merck Index:13,01231
- Melting Pt:232…233 °C
- Density:1.41 g/cm³ (20 °C)
- Storage Temperature:Refrigerator
- EINECS:200-399-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



