Order Entry
Export
Orders LinkContactUsLinkComponent
 

Coronene 95%

Irritant
Coronene 95%
Certificates
Coronene 95%

2 Options Available Below

Product Details & Documents
Synonyms:  Hexabenzobenzene
  • CAS Number:191-07-1
  • Formula:C₂₄H₁₂
  • MDL Number:MFCD00004134
  • Molecular Weight:300.36 g/mol
  • Boiling Pt:525 °C (1013 hPa)
  • Melting Pt:438…440 °C
  • Density:1,371 g/cm³ (25 °C)
  • EINECS:205-881-7
  • UN:2811

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    Yellow to green Crystalline powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥94 %
  • Titration RCOCl:
    ≥96.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 46,90 €
    46,90 €
    1 * 100 mg
       46,90 €

  • Glass bottle for solids
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 153,00 €
    153,00 €
    1 * 500 mg
       153,00 €