7-Methylindole
Glass bottle for solids, 100 g
Catalog # COBBIN-0033-100G
Supplier: COMBI-BLOCKS
Jump to:
Product Details & Documents
- CAS Number:933-67-5
- Formula:C₉H₉N
- MDL Number:MFCD00005684
- Molecular Weight:131.18 g/mol
- Boiling Pt:266 °C (1013 hPa)
- Melting Pt:83 °C
- Density:1,0202 g/cm³ (99.6 °C)
- :Density: 1.0202 g/cm³ (99.6 °C)
- EINECS:213-270-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



