5-Nitroisoquinoline
Glass bottle for solids, 100 g
Catalog # COBBAN-1114-100G
Supplier: COMBI-BLOCKS
Jump to:
Product Details & Documents
- CAS Number:607-32-9
- Formula:C₉H₆N₂O₂
- MDL Number:MFCD00006905
- Molecular Weight:174.16 g/mol
- Boiling Pt:340 °C (1013 hPa)
- Melting Pt:111…113 °C
- Storage Temperature:Ambient
- EINECS:210-133-8
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



