0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
5,6-Dihydroxyflavone
5,6-Dihydroxyflavone ≥97%
5,6-Dihydroxyflavone ≥97%
Irritant
5,6-Dihydroxyflavone ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
5,6-Dihydroxyflavone ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
6665-66-3
:
Formula:
C₁₅H₁₀O₄
MDL Number:
MFCD00601534
:
254.24 g/mol
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH27039
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation