Order Entry
Export
Orders LinkContactUsLinkComponent
 

4-Hydroxyindole

Glass bottle for solids, 25 g
Catalog # COBBIN-0034-25G
Irritant
Supplier:   COMBI-BLOCKS
4-Hydroxyindole
Catalog # COBBIN-0034-25G
Supplier:  COMBI-BLOCKS
CAS Number:  2380-94-1
4-Hydroxyindole

Glass bottle for solids, 25 g

CAS Number:  2380-94-1
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Indol-4-ol, 4-Indolol, 4-Hydroxy-1H-indole, 1H-Indol-4-ol
  • CAS Number:2380-94-1
  • Formula:C₈H₇NO
  • MDL Number:MFCD00005667
  • Molecular Weight:133.15 g/mol
  • Boiling Pt:100 °C (0,01 mmHg)
  • Boiling Pt:100 °C (0.01 mmHg)
  • Melting Pt:102…104 °C
  • EINECS:219-177-2

 

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications