Daidzein
Glass bottle for solids, 25 mg
Catalog # ROTH6047.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Daidzeol, 7-Hydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 7-Hydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',7-Dihydroxyisoflavone, 7-Hydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one
- CAS Number:486-66-8
- Formula:C₁₅H₁₀O₄
- MDL Number:MFCD00016954
- Molecular Weight:254.24 g/mol
- Boiling Pt:513 °C (1013 hPa)
- Melting Pt:212…214 °C
- EINECS:207-635-4
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 mg



