Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Cyanobenzenesulphonamide
4-Cyanobenzenesulphonamide 97%
4-Cyanobenzenesulphonamide 97%
:
Thermo Fisher Scientific
4-Cyanobenzenesulphonamide 97%
:
Thermo Fisher Scientific
:
3119-02-6
:
:
C₇H₆N₂O₂S
:
MFCD00178767
:
182.2 g/mol
:
168…171 °C
:
221-492-5
:
GHS07
:
Ambient
msds_ALFAH31836
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years