Order Entry
Export
Orders LinkContactUsLinkComponent
4-Bromo-2-fluoropyridine

4-Bromo-2-fluoropyridine

Glass bottle for solids, 25 g
Catalog # COBBOS-7555-25G
Irritant
Supplier:   COMBI-BLOCKS
4-Bromo-2-fluoropyridine
Catalog # COBBOS-7555-25G
Supplier:  COMBI-BLOCKS
CAS Number:  128071-98-7
4-Bromo-2-fluoropyridine

Glass bottle for solids, 25 g

CAS Number:  128071-98-7
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:128071-98-7
  • Formula:C₅H₃BrFN
  • MDL Number:MFCD04112504
  • Molecular Weight:175.99 g/mol
  • UN:2810
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications