4-Bromo-2,6-difluorobenzaldehyde, Sigma-Aldrich®
Catalog # SIAL706957-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:221 g/mol
CAS Number:537013-51-7
Formula:C₇H₃BrF₂O
MDL Number:MFCD03094459