Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-1-methylimidazole
4-Bromo-1-methylimidazole, Sigma-Aldrich®
4-Bromo-1-methylimidazole, Sigma-Aldrich®
4-Bromo-1-methylimidazole, Sigma-Aldrich®
SIAL711292-1G
:
Merck
:
4-Bromo-1-methylimidazole, Sigma-Aldrich®
SIAL711292-1G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
161 g/mol
:
25676-75-9
:
C₄H₅BrN₂
:
MFCD01320501