4,4'-Diaminobenzanilide
Catalog # MOLE16774468-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:227.27 g/mol
Melting Pt:214…215 °C
EINECS:212-321-5
CAS Number:785-30-8
Formula:C₁₃H₁₃N₃O
MDL Number:MFCD00025361