Order Entry
Export
Orders LinkContactUsLinkComponent
Luteolin

Luteolin

Glass bottle for solids, 25 mg
Catalog # MOLEM46263693
Irritant
Supplier:   Molekula
Luteolin
Catalog # MOLEM46263693
Supplier:  Molekula
CAS Number:  491-70-3
Luteolin

Glass bottle for solids, 25 mg

CAS Number:  491-70-3
Price
Inquire For Price
Log in for availability
1 * 25 mg
Product Details & Documents
Synonyms:  2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-4-benzopyrone, Natural Yellow 2, 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-4H-chromen-4-one, 3',4',5,7-Tetrahydroxyflavone
  • CAS Number:491-70-3
  • Formula:C₁₅H₁₀O₆
  • MDL Number:MFCD00017309
  • Molecular Weight:286.24 g/mol
  • EINECS:207-741-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg

Specifications