Luteolin
Glass bottle for solids, 10 mg
Catalog # ROTH4546.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-4-benzopyrone, Natural Yellow 2, 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-4H-chromen-4-one, 3',4',5,7-Tetrahydroxyflavone
- CAS Number:491-70-3
- Formula:C₁₅H₁₀O₆
- MDL Number:MFCD00017309
- Molecular Weight:286.24 g/mol
- EINECS:207-741-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg



