Order Entry
Export
Orders LinkContactUsLinkComponent
Luteolin ≥97%

Luteolin ≥97%

Luteolin ≥97%
Certificates
Luteolin ≥97%

2 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 491-70-3 Molecular Formula: C15H10O6 Formula Weight: 286.24 Melting Point: 300° Beilstein Reference: 292084 Merck Reference: 14,5614 MDL No.: MFCD00017309

Synonyms:  2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-4-benzopyrone, Natural Yellow 2, 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-4H-chromen-4-one, 3',4',5,7-Tetrahydroxyflavone
  • CAS Number:491-70-3
  • Formula:C₁₅H₁₀O₆
  • MDL Number:MFCD00017309
  • Molecular Weight:286.24 g/mol
  • Merck Index:14,5614
  • Melting Pt:>300 °C
  • Storage Temperature:Ambient
  • EINECS:207-741-0
Product Family Options

Product Information