
4,6-Decadiyne ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16387-71-6
:
Formula:C₁₀H₁₄
MDL Number:MFCD00041645
Boiling Pt:96 °C (15 mmHg)
Density:0.82
UN:3295
ADR:3,III
:134.22 g/mol
:GHS07
Storage Temperature:Ambient