4,5-Dicyanoimidazole, Sigma-Aldrich®
Catalog # SIAL324132-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:118.1 g/mol
Melting Pt:176 °C
Density:1.458 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:214-344-6
CAS Number:1122-28-7
Formula:C₅H₂N₄
MDL Number:MFCD00005194