

4,5-Dicyanoimidazole ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1122-28-7
:
Formula:C₅H₂N₄
MDL Number:MFCD00005194
:118.1 g/mol
Melting Pt:173…177 °C
Density:1.458 g/cm³ (20 °C)
EINECS:214-344-6
:GHS05
:GHS07
Storage Temperature:Ambient