4,5-Dichloroimidazole, Sigma-Aldrich®
Catalog # SIAL277363-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:136.97 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:15965-30-7
Formula:C₃H₂Cl₂N₂
MDL Number:MFCD00005195