Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Dichloro-2-methylimidazole
4,5-Dichloro-2-methylimidazole
4,5-Dichloro-2-methylimidazole
4,5-Dichloro-2-methylimidazole
Supplier:
Thermo Scientific
Certificates
Expand image
4,5-Dichloro-2-methylimidazole
Supplier:
Thermo Scientific
About this item
CAS Number:
15965-33-0
:
Formula:
C₄H₄Cl₂N₂
MDL Number:
MFCD00051639
:
150.99 g/mol