3,5-Dinitrobenzamide
Catalog # EHERC15588500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
Melting Pt:183 °C
:211.13 g/mol
EINECS:204-499-8
CAS Number:121-81-3
Formula:C₇H₅N₃O₅
MDL Number:MFCD00007985