Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dimethoxybenzamide
3,5-Dimethoxybenzamide
3,5-Dimethoxybenzamide
3,5-Dimethoxybenzamide
Supplier:
CHEMPUR
Certificates
Expand image
3,5-Dimethoxybenzamide
Supplier:
CHEMPUR
About this item
CAS Number:
17213-58-0
:
Formula:
C₉H₁₁NO₃
MDL Number:
MFCD00007987
:
181.19 g/mol
Boiling Pt:
345 °C (1013 hPa)
Melting Pt:
148 °C
EINECS:
241-257-0