3,6-Dihydroxyflavone ≥97%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
3,6-Dihydroxyflavone ≥97%
Catalog # H26989.06
Supplier: Thermo Fisher Scientific
CAS Number: 108238-41-1
Jump to:
Product Details & Documents
Purity: 97% CAS No.: 108238-41-1 Molecular Formula: C15H10O4 Formula Weight: 254.24 Melting Point: 231-234° MDL No.: MFCD00143082
Synonyms: 3,6-Dihydroxy-2-phenyl-4-benzopyrone, 3,6-Dihydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 3,6-Dihydroxy-2-phenyl-4H-chromen-4-one, 3,6-Dihydroxy-2-phenyl-4H-1-benzopyran-4-one, 3,6-Dihydroxy-2-phenylchromen-4-one
- CAS Number:108238-41-1
- Formula:C₁₅H₁₀O₄
- MDL Number:MFCD00143082
- Molecular Weight:254.24 g/mol
- Melting Pt:231…234 °C
- Storage Temperature:Ambient
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



