Order Entry
Export
Orders LinkContactUsLinkComponent
3,5-Dichlorosalicylaldehyde

3,5-Dichlorosalicylaldehyde

Supplier:   Thermo Scientific
3,5-Dichlorosalicylaldehyde
Supplier:  Thermo Scientific
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  3,5-Dichloro-2-hydroxybenzaldehyde
  • CAS Number:90-60-8
  • Formula:C₇H₄Cl₂O₂
  • MDL Number:MFCD00003320
  • Molecular Weight:191.01 g/mol
  • Boiling Pt:723 °C (1013 hPa)
  • Melting Pt:92 °C
  • EINECS:202-005-5
Specifications for Product Family

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

    Specifications:
    More
    Specifications
    Cat. No.
    MAYBSB01828.1
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

    Specifications:
    More
    Specifications
    Cat. No.
    MAYBSB01828EA
  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g

    Specifications:
    More
    Specifications
    Cat. No.
    MAYBSB01828.50