
3,4-Dihydroxy-3-cyclobutene-1,2-dione ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2892-51-5
:
Formula:C₄H₂O₄
MDL Number:MFCD00001334
:114.06 g/mol
Melting Pt:>300 °C
Density:1 g/cm³ (20 °C)
Flash Pt:190 °C (374 °F)
Storage Temperature:Ambient
EINECS:220-761-4
UN:3261
ADR:8,II
:GHS07