3,4-Dihydroxy-3-cyclobutene-1,2-dione
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:2892-51-5
:
Formula:C₄H₂O₄
MDL Number:MFCD00001334
:114.06 g/mol
Melting Pt:293 °C
Density:1 g/cm³ (20 °C)
EINECS:220-761-4
UN:3261
ADR:8,II