Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 3,4-diaminobenzoate
Methyl-3,4-diaminobenzoate 98%
Methyl-3,4-diaminobenzoate 98%
:
Thermo Fisher Scientific
Methyl-3,4-diaminobenzoate 98%
:
Thermo Fisher Scientific
:
36692-49-6
:
:
C₈H₁₀N₂O₂
:
MFCD00017098
:
319 °C (1013 hPa)
:
105…108 °C
:
166.18 g/mol
:
GHS07
:
Ambient
msds_ALFAA14656
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years