Order Entry
Export
Orders LinkContactUsLinkComponent
Naphthol AS acetate

Naphthol AS acetate

Glass bottle for solids, 5 g
Catalog # MOLE25134487-5G
Supplier:   Molekula
Naphthol AS acetate
Catalog # MOLE25134487-5G
Supplier:  Molekula
CAS Number:  1163-67-3
Naphthol AS acetate

Glass bottle for solids, 5 g

CAS Number:  1163-67-3
Price
225,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  2-Hydroxy-3-naphthoic acid Anilide Acetate, 2-(N-Phenylcarbamoyl)-3-naphthyl acetate, 2-Acetoxy-3-naphthoic acid Anilide, Naphthol AS Acetate, 3-Acetoxy-2-naphthanilide, Acetyl Naphthol AS
  • CAS Number:1163-67-3
  • Formula:C₁₉H₁₅NO₃
  • MDL Number:MFCD00004095
  • Molecular Weight:305.33 g/mol
  • EINECS:214-608-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications